Anti-CFTR Rabbit Recombinant Antibody [clone: CFTR/2290R]
Catalog # ANTIA248162-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:ABC 35
- Antigen symbol:CFTR
- Clonality:Recombinant
- Clone:CFTR/2290R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Form:Liquid
- Gene ID:UniprotID# P13569
- Antigen synonyms:ABC35|ABCC7|ATP-binding cassette sub-family C member 7|cAMP-dependent chloride channel|cAMP dependent chloride channel|ATP binding cassette transporter sub family C member 7|ABCC 7|ATP binding cassette sub family C member 7|ATP Binding Cassette Superfamily C Member 7
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:165 - 170 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human CFTR protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [CFTR/2290R] antibody to CFTR for IHC-P with samples derived from human and mouse.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CFTR
Clonality: Recombinant
Clone: CFTR/2290R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse