Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD52 Rabbit Recombinant Antibody [clone: CD52/2276R]
Anti-CD52 Rabbit Recombinant Antibody [clone: CD52/2276R]
Catalog # ANTIA248109-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD52 Rabbit Recombinant Antibody [clone: CD52/2276R]
Catalog # ANTIA248109-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cambridge pathology 1 antigen
  • Antigen symbol:
    CD52
  • Clonality:
    Recombinant
  • Clone:
    CD52/2276R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Monkey,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P31358
  • Antigen synonyms:
    CD 52|CD52 antigen|CAMPATH 1|CD52_HUMAN|CAMPATH-1 antigen|CD52|CDw52|CD52 molecule|CAMPATH 1 antigen
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    6 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human CD52 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [CD52/2276R] antibody to CD52 for IHC-P with samples derived from human and monkey.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD52
Clonality: Recombinant
Clone: CD52/2276R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey