Anti-TRPV3 Mouse Monoclonal Antibody [clone: N15/39]
Catalog # ANTIA305015-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:1110036I10Rik
- Antigen symbol:TRPV3
- Clonality:Monoclonal
- Clone:N15/39
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q4QYD9
- Antigen synonyms:TRPV3|Vanilloid receptor like 3|MGC124324|Vanilloid receptor 3|Trpv3 heat sensitive channel|Vanilloid receptor-like 3|TRPV3_HUMAN|MGC124325|Transient receptor potential cation channel subfamily V member 3
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
- Molecular weight:~70 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide amino acids 458 - 474 (cytoplasmic C - terminus) of rat TrpV3.
- Tested applications:IHC, ICC/IF
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [N15/39] antibody to TRPV3 for WB, IHC, ICC/IF, IP and antibody microarray with samples derived from human, mouse and rat.
- Validated applications: WB, IHC, ICC/IF, IP, Antibody microarray
- Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100
Type: Primary
Antigen: TRPV3
Clonality: Monoclonal
Clone: N15/39
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Mouse;Rat