Anti-SHANK3 Mouse Monoclonal Antibody [clone: S367-51]
Catalog # ANTIA304719-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:AI841104
- Antigen symbol:SHANK3
- Clonality:Monoclonal
- Clone:S367-51
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9JLU4
- Antigen synonyms:Proline rich synapse associated protein 2|SH3/ankyrin domain gene 3|SH3 and multiple ankyrin repeat domains 3|DEL22q13.3|PSAP2|Proline-rich synapse-associated protein 2|KIAA1650|ProSAP2|SH3 and multiple ankyrin repeat domains protein 3
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,1% sodium azid
- Molecular weight:~190 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein amino acids 538 - 626 (SH3 domain) of rat SHANK3. Mouse: 100% identity (89/89 amino acids identical). Human: 97% identity (87/89 amino acids identical). ~70% identity with SHANK1 and SHANK2..
- Tested applications:IHC, ICC/IF
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [S367-51] antibody to SHANK3 for WB, IHC and ICC/IF with samples derived from human, mouse and rat.
- Validated applications: WB, IHC, ICC/IF
- Recommended dilutions: WB: 1:1000, ICC/IF: 1:100
Type: Primary
Antigen: SHANK3
Clonality: Monoclonal
Clone: S367-51
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Mouse;Rat