Anti-HSP70 Rabbit Recombinant Antibody [clone: RM432]
Catalog # ANTIA290851-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:DnaK type molecular chaperone HSP70 1
- Antigen symbol:HSP70
- Clonality:Recombinant
- Clone:RM432
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P0DMV8, P0DMV9
- Antigen synonyms:Heat shock 70 kDa protein 1|FLJ54303|FLJ75127|FLJ54408|Epididymis secretory protein Li 103|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|FLJ54370|FLJ54392
- Storage buffer:Supplied in phosphate buffered saline with 50% glycerol, 1% BSA, and 0,09% sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human HSP70 protein.
- Tested applications:IHC
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Recombinant rabbit monoclonal [RM432] antibody to HSP70 for IHC and WB with samples derived from human.
- Validated applications: IHC, WB
- Recommended dilutions: IHC: 1:100 to 1:500, WB: 1:2000 to 1:5000
Type: Primary
Antigen: HSP70
Clonality: Recombinant
Clone: RM432
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human