Anti-CLEC7A Mouse Monoclonal Antibody [clone: 15E2] (PE (Phycoerythrin))
Catalog # ANTIA285836-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Beta GR
- Antigen symbol:CLEC7A
- Clonality:Monoclonal
- Clone:15E2
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Epitope:LWEDGSTFSSN
- Form:Liquid
- Gene ID:UniprotID# Q9BXN2
- Antigen synonyms:CANDF4|C type lectin domain family 7 member a|C type calcium dependent carbohydrate recognition domain lectin superfamily member 12|BGR|C-type lectin domain family 7 member A|CLC7A_HUMAN|Beta-glucan receptor|C-type lectin superfamily member 12|CLEC7A
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Avoid prolonged exposure to light. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Extracellular part of human CD369 with hIgG FC tag.
- Tested applications:IHC-Fr
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [15E2] antibody to CLEC7A (PE) for Flow cytometry and IHC-Fr with samples derived from human.
- Validated applications: Flow cytometry, IHC to Fr
- Recommended dilutions: Flow cytometry: The reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CLEC7A
Clonality: Monoclonal
Clone: 15E2
Conjugation: PE (Phycoerythrin)
Epitope: LWEDGSTFSSN
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human