Anti-CD59 Rabbit Recombinant Antibody [clone: MACIF/7021R]
Catalog # ANTIA278689-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:16.3A5
- Antigen symbol:CD59
- Clonality:Recombinant
- Clone:MACIF/7021R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P13987
- Antigen synonyms:CD59 antigen|1F5|CD59 antigen complement regulatory protein |CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, EJ30, EL32 and G344)|CD 59|CD59|20 kDa homologous restriction factor|CD59 antigen p18 20|1F5 antigen
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:20 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full length human CD59 protein.
- Tested applications:IHC
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [MACIF/7021R] antibody to CD59 for IHC with samples derived from human.
- Validated applications: IHC
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD59
Clonality: Recombinant
Clone: MACIF/7021R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human