Anti-Testosterone Rabbit Recombinant Antibody [Clone: RM435]
Catalog # ANTIA290880-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:17b hydroxy 4 androsten 3 one
- Antigen symbol:Testosterone
- Clonality:Recombinant
- Clone:RM435
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Testosterone|17ß-hydroxyandrost-4-ene-3-one
- Storage buffer:Supplied in phosphate buffered saline with 50% glycerol, 1% BSA, and 0,09% sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Testosterone - KLH.
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Recombinant rabbit monoclonal [RM435] antibody to testosterone for ELISA.
- Validated applications: ELISA
- Recommended dilutions: cELISA: 1 to 10 ng/well
Type: Primary
Antigen: Testosterone
Clonality: Recombinant
Clone: RM435
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: