Order Entry
Netherlands
ContactUsLinkComponent
Anti-C5AR2 Mouse Monoclonal Antibody [Clone: 1D9-M12] (PE (Phycoerythrin))
Anti-C5AR2 Mouse Monoclonal Antibody [Clone: 1D9-M12] (PE (Phycoerythrin))
Catalog # ANTIA285833-100
Supplier:  Antibodies.com
New Product
undefined
Anti-C5AR2 Mouse Monoclonal Antibody [Clone: 1D9-M12] (PE (Phycoerythrin))
Catalog # ANTIA285833-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C5a anaphylatoxin chemotactic receptor C5L2
  • Antigen symbol:
    C5AR2
  • Clonality:
    Monoclonal
  • Clone:
    1D9-M12
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9P296
  • Antigen synonyms:
    G protein coupled receptor C5L2|GPR77|G protein coupled receptor 77|GPCR C5L2|G-protein coupled receptor 77|E030029A11Rik|GPF77|C5ARL_HUMAN
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Avoid prolonged exposure to light. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    L1.2 cells transfected with human C5aR2.
  • Purification:
    This antibody is conjugated with PE under optimum conditions The conjugate is purified by size-exclusion chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [1D9-M12] antibody to C5AR2 (PE) for Flow cytometry with samples derived from human and non-human primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: The reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: C5AR2
Clonality: Monoclonal
Clone: 1D9-M12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Non-Human Primates