Bestelling invoeren
Netherlands
ContactUsLinkComponent
Monkey Osteocalcin ELISA Kit
Monkey Osteocalcin ELISA Kit
Catalog # ANTIA246963-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Monkey Osteocalcin ELISA Kit
Catalog # ANTIA246963-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • VPE:
    96 T
  • Duur van de test:
    Multiple steps
  • Assaytype:
    Sandwich
  • Formaat:
    Pre-coated
  • Primary antibody reactivity:
    Monkey
  • Target protein:
    Osteocalcin
  • Omschrijving:
    Monkey osteocalcin ELISA kit
  • Monstertype:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Kruisreactiviteit:
    Monkey Osteocalcin ELISA Kit exhibits high specificity and excellent specificity for the detection of monkey Osteocalcin. No significant cross-reactivity or interference between Osteocalcin and analogues was observed.
  • Detectiemethode:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Houdbaarheid:
    Store for 6 months at 4 °C
  • Detectiebereik:
    0,625 - 40 ng/ml
  • Opslagtemperatuur:
    4 °C
  • Monstervolume:
    100 μl
  • Gevoeligheid:
    0,375 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Monkey Osteocalcin ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of monkey Osteocalcin in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Monkey Osteocalcin ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of monkey Osteocalcin in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Osteocalcin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Osteocalcin present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Osteocalcin Antibody, which binds the captured Osteocalcin present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Osteocalcin captured in each well. The concentration of Osteocalcin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.