Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Mouse IL-16 ELISA Kit

Mouse IL-16 ELISA Kit

Mouse IL-16 ELISA kit, 96 Tests
Catalog # ANTIA310669-96
Supplier:   Antibodies.com
Mouse IL-16 ELISA Kit
Catalog # ANTIA310669-96
Supplier:  Antibodies.com
CAS Number:  
Mouse IL-16 ELISA Kit
Mouse IL-16 ELISA Kit

Mouse IL-16 ELISA kit, 96 Tests

CAS Number:  
Price
565,00 €
Log in for availability
1 * 1 EA
Product Details & Documents

Mouse IL-16 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse IL-16 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 15 to 240 ng/L
  • Sensitivity: 0,31 ng/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse IL-16 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse IL-16 in serum, plasma or other biological fluids. An antibody specific for IL-16 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IL-16 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-IL-16 antibody, which also binds the IL-16 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-IL-16 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-IL-16 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of IL-16 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of IL-16 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Mouse IL-16 ELISA kit
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Serum, plasma or other biological fluids
  • Sample volume:
    40 μl
  • Detection range:
    15 - 240 ng/L
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ANTIA310669-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,31 ng/L
  • Format:
    Pre-coated
  • Target protein:
    IL-16
  • Regulatory status:
    RUO

Specifications