Order Entry
Netherlands
ContactUsLinkComponent
Human MMS21 ELISA Kit
Human MMS21 ELISA Kit
  ANTIA310419-96
New Product
 :  
undefined
Human MMS21 ELISA Kit
  ANTIA310419-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    MMS21
  • Description:
    Human MMS21 ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    125 - 2000 ng/L
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    10,12 ng/L
  • Regulatory status:
    RUO

 

 

Human MMS21 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human MMS21 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 125 to 2000 ng/L
  • Sensitivity: 10,12 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human MMS21 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human MMS21 in serum, plasma or other biological fluids. An antibody specific for MMS21 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the MMS21 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-MMS21 antibody, which also binds the MMS21 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-MMS21 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-MMS21 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of MMS21 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of MMS21 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.