Order Entry
Netherlands
ContactUsLinkComponent
Rat PACAP-38 ELISA Kit
Rat PACAP-38 ELISA Kit
Catalog # ANTIA74924-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Rat PACAP-38 ELISA Kit
Catalog # ANTIA74924-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    PACAP-38
  • Description:
    Rat PACAP-38 ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Rat PACAP-38 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat PACAP-38. No significant cross-reactivity or interference between PACAP-38 and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    125 - 8000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    75 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Rat PACAP-38 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat PACAP-38 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection range: 125 to 8000 pg/ml
  • Sensitivity: 75 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%

Rat PACAP-38 ELISA Kit (A74924) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat PACAP-38 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PACAP-38 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PACAP-38 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PACAP-38 Antibody, which binds the captured PACAP-38 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PACAP-38 captured in each well. The concentration of PACAP-38 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.