Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Competitive
- Format:Pre-coated
- Host:
- Primary antibody reactivity:Mouse
- Target protein:Dehydroepiandrosterone
- Description:Mouse dehydroepiandrosterone ELISA kit
- Environmentally Preferable:
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Mouse Dehydroepiandrosterone ELISA kit exhibits high specificity and excellent specificity for the detection of mouse Dehydroepiandrosterone. No significant cross-reactivity or interference between Dehydroepiandrosterone and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,156 - 10 ng/ml
- Storage temperature:4 °C
- Sample volume:50 μl
- Sensitivity:0,094 ng/ml
- Regulatory status:RUO
Specifications
About this item
Mouse Dehydroepiandrosterone ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse Dehydroepiandrosterone in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 0,156 to 10 ng/ml
- Sensitivity: 0,094 ng/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse Dehydroepiandrosterone ELISA kit (A76438) employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse Dehydroepiandrosterone in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Dehydroepiandrosterone antigen. During the incubation, Dehydroepiandrosterone present in the samples or standards competes with the fixed amount of immobilised Dehydroepiandrosterone for binding sites on the Biotinylated Anti-Dehydroepiandrosterone Antibody. The more Dehydroepiandrosterone present in a sample or standard, the less Biotinylated Anti-Dehydroepiandrosterone Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Dehydroepiandrosterone Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Dehydroepiandrosterone present in each sample or standard. The concentration of Dehydroepiandrosterone can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.