Order Entry
Netherlands
ContactUsLinkComponent
Magicard™ Colour Film for Pronto 100, 200 Shot
Magicard™ Colour Film for Pronto 100, 200 Shot
Catalog # BRDY321976
Supplier:  Brady
New Product
undefined
Magicard™ Colour Film for Pronto 100, 200 Shot
Catalog # BRDY321976
Supplier:  Brady
New Product

Specifications

  • Colour:
    Assorted
  • Description:
    200 shot colour film, 4-colour ribbon (Cyan, Magenta, Yellow, Black), 200 single-sided cards
  • Length:
    10 m
  • Pk:
    1
  • Width:
    60 mm
  • For:
    Pronto 100

Specifications

About this item

The Magicard™ colour ribbon is perfect for full colour prints with a Pronto 100 printer.

  • Overlay panel for extra print protections
  • Print 200 cards in one go

This high-quality, YMCKO ribbon guarantees to print 200 cards on the Pronto 100 printer.

Delivery: Includes 1 dye film and 1 cleaning roller.