- Antigen name:TRA-1-81
- Clonality:Monoclonal
- Clone:TRA-1-81
- Host:Mouse
- Isotype:IgM kappa
- Reactivity:Rhesus monkey,Human,Rat
- Gene ID:5420
- Pk:100 µG
Mouse monoclonal IgM antibody against human, rat, rhesus TRA-1-81, unconjugated.
- Pluripotent stem cells
This monoclonal antibody reacts with a carbohydrate epitope on TRA-1-81, a high-molecular-mass (> 200 kDa) glycoprotein expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), embryonic stem (ES), and induced pluripotent stem (iPS) cells, as well as rhesus monkey and rat ES cell lines. No immunoreactivity is observed with mouse EC, EG, ES, or iPS cells. The epitope of the TRA-1-81 antibody is associated with a keratan-sulfated transmembrane protein identified as podocalyxin and is resistant to treatment with neuraminidase.
Type: Anti-TRA-1-81 Mouse Monoclonal Antibody [clone: TRA-1-81]
Antigen: TRA-1-81
Clonality: Monoclonal
Clone: TRA-1-81
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human, Rat, Rhesus monkey