Order Entry
Netherlands
ContactUsLinkComponent
 
Human Recombinant IFN-alpha 2B
 
undefined
Human Recombinant IFN-alpha 2B

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    IFN-alpha 2B
  • Purity:
    ≥95%

 

Frequently Bought Together