Influenza (HLA Class I Control) Peptide Pool
Catalog # STMC100-0672
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:1 KIT
- Source:
- Species:
- Seq:0075
- Environmentally Preferable:
- Protein/peptide name:Influenza (HLA Class I Control) Peptide Pool
- Purity:≥95%
Specifications
About this item
Influenza (HLA class I control) peptide pool is a lyophilised mixture of 17 peptides from the influenza virus. The pool consists of defined HLA class I-restricted T cell epitopes, and can be used as an antigen-specific positive control to stimulate T cells.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').