HPV18 (L1) Peptide Pool
Catalog # STMC100-1396
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:1 AU
- Protein/peptide name:HPV18 (L1) Peptide Pool
- Purity:Average 70%
Specifications
About this item
Stimulating T cells with the HPV18 (L1) Peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. A lyophilised mixture of 140 peptides from major capsid protein L1 of human papillomavirus type 18 (HPV18), HPV18 (L1) Peptide pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 568 on L1. L1 enters the nuclei of host cells and can assemble into virus-like particles (Stauffer et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').