Order Entry
Netherlands
ContactUsLinkComponent
MBD
MBD
Catalog # M5760-1G
CAS Number:  
undefined
MBD
Catalog # M5760-1G
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance (Colour):
    Orange to Red
  • Appearance (Form):
    Powder
  • Proton NMR spectrum:
    Conforms to Structure
  • Carbon:
    54.3 - 57.7 %
  • Nitrogen:
    18.1 - 19.3 %
  • Purity (HPLC):
    ≥97.5 %
  • Solubility (Colour):
    Orange
  • Solubility (Turbidity)
    50 mg/ml DMSO:CH3OH (1:1):
    Clear

Specification Test Results

About this item

 :
 :300.274 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
CAS Number:33984-50-8
Formula:C₁₄H₁₂N₄O₄
MDL Number:MFCD00042778