SignalStain® Proliferation/Apoptosis IHC Sampler Kit
Catalog # CSTA8109S
Supplier: Cell Signaling
Specifications
- ImmunoChemistry:Yes
- Storage temperature:+4 °C and −20 °C
- Shipping temperature:Cold
- Pk:1 KIT
Specifications
About this item
The SignalStain® Proliferation/Apoptosis IHC Sampler Kit from Cell Signalling Technology allows the researcher to examine paraffin-embedded tissues or cells with antibodies that will detect cellular apoptosis or proliferation.
Each antibody is validated for use in immunohistochemical assays using multiple approaches. Also included in the kit are control slides that can be used to verify the performance of each antibody and a primary antibody diluent. Please see table above for recommended diluent for each antibody in this kit.
Each antibody in the SignalStain® Proliferation/Apoptosis IHC Sampler Kit detects endogenous levels of its target protein and does not cross-react with any related proteins.
Delivery: Kit includes Cleaved Caspase-3 (Asp175) Antibody (CSTA9661) 40 µl, Phospho-Histone H3 (Ser10) Antibody (CSTA9701) 40 µl, PCNA (PC10) Mouse mAb (CSTA2586) 40 µl, Survivin (71G4B7) Rabbit mAb (CSTA2808) 40 µl, SignalStain® Antibody Diluent (CSTA8112) 25 ml, SignalSlide® Cleaved Caspase-3 (Asp175) IHC Controls (CSTA8104) 1 pack.