Anti-NTHL1 Rabbit Polyclonal Antibody
PRSI55-909
New Product
- Type antilichaam:Primair
- antigeen symbool:NTHL1
- Clonality:Polyklonaal
- Conjugatie:Onconjugeerd
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitoop:Amino Acid Range = 88-117
- Formulier:Liquid
- Gen-ID:P78549
- Antigeen synoniemen:NTHL1
- Modification:None
- Opslagbuffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Moleculair gewicht:34 kDa
- Opslagtemperatuur:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentratie:0,35 mg/ml
- Shipping temperature:4 °C
- Immunogeen:This NTHL1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 88-117 amino acids from the Central region of human NTHL1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Maat:400 µl
- VPE:400 µl
The protein encoded by this gene is a DNA N-glycosylase of the endonuclease III family. Like a similar protein in E. coli, the encoded protein has DNA glycosylase activity on DNA substrates containing oxidized pyrimidine residues and has apurinic/apyrimidinic lyase activity.
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: NTHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human