Order Entry
Netherlands
ContactUsLinkComponent
Anti-INSR Recombinant Antibody [Clone: 83-14]
Anti-INSR Recombinant Antibody [Clone: 83-14]
Catalog # PRSI24-173
Supplier:  ProSci Inc.
Anti-INSR Recombinant Antibody [Clone: 83-14]
Catalog # PRSI24-173
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    insulin receptor
  • Antigen symbol:
    INSR
  • Clonality:
    Recombinant
  • Clone:
    83-14
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Pig,
    Sheep,
    Human,
    Bovine
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    3643
  • Antigen synonyms:
    IR|HHF5|Insulin Receptor alpha|CD220|HIR
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    This antibody was raised by immunizing mice with IM-9 lymphocytes followed by purified insulin receptor.
  • Size:
    0,2 mg
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: insulin receptor
Clonality: Recombinant
Clone: 83-14
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2a kappa
Reactivity: Human;Bovine;Pig;Sheep