Anti-MS4A1 Recombinant Antibody [Clone: 10F381 (rituximab)]
PRSI24-125
New Product
- Antibody type:Primary
- Antigen name:membrane spanning 4-domains A1
- Antigen symbol:MS4A1
- Clonality:Recombinant
- Clone:10F381 (rituximab)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Isotype:IgG1 kappa
- Reactivity:Cynomolgus monkey,Rhesus monkey,Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:931
- Antigen synonyms:B-lymphocyte surface antigen B1|Membrane-spanning 4-domains subfamily A member 1|Bp35 Leukocyte surface antigen Leu-16|CD20|Leu16|B-lymphocyte antigen CD20
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Human lymphoblastoid cell line SB.
- Size:0,2 mg
- Pack type:Vial
- Pk:0,2 mg
Type: Primary,Biosimilars
Antigen: membrane spanning 4-domains A1
Clonality: Recombinant
Clone: 10F381 (rituximab)
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human;Rhesus Monkey;Cynomolgus Monkey