Anti-Fcgr2b Recombinant Antibody [Clone: 2.4G2]
PRSI24-121
New Product
- Antibody type:Primary
- Antigen name:Fc receptor, IgG, low affinity Iib
- Antigen symbol:Fcgr2b
- Clonality:Recombinant
- Clone:2.4G2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Mouse
- Cross adsorption:No
- Form:Liquid
- Gene ID:14130
- Antigen synonyms:Fcγ III/II Receptor|IgG Fc receptor III|Fc-gamma RIII|Low affinity immunoglobulin gamma Fc region receptor II|CD16|Fc gamma receptor IIB|FcRII IgG Fc rec|Fc-gamma RII|Low affinity immunoglobulin gamma Fc region receptor III|FcRIII|Fc-gamma-RIIB
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Mouse BALB/c Macrophage J774 Cell Line.
- Tested applications:Blocking
- Size:0,2 mg
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: Fc receptor, IgG, low affinity Iib
Clonality: Recombinant
Clone: 2.4G2
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Mouse