Order Entry
Netherlands
ContactUsLinkComponent
Anti-CDH16 Mouse Monoclonal Antibody [clone: CDH16/1071]
Anti-CDH16 Mouse Monoclonal Antibody [clone: CDH16/1071]
Catalogus # PRSI33-309
Supplier:  ProSci Inc.
New Product
Anti-CDH16 Mouse Monoclonal Antibody [clone: CDH16/1071]
Catalogus # PRSI33-309
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    cadherin 16, KSP-cadherin
  • Antigen symbol:
    CDH16
  • Clonality:
    Monoclonal
  • Clone:
    CDH16/1071
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    O75309
  • Antigen synonyms:
    CDH16
  • Storage buffer:
    PBS with 0,1 mg/ml BSA and 0,05% sodium azide
  • Storage temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0,2 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    Recombinant human protein was used as the immunogen for the Cadherin 16 antibody.
  • Purification:
    Protein G affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

This mAb recognises a protein of 130 kDa, identified as Ksp-cadherin. Cadherins form a superfamily of related glycoproteins that mediate calcium-dependent cell adhesion and transmit signals from the extracellular matrix to the cytoplasm. Cadherins have been implicated in embryogenesis, tissue morphogenesis, tissue structure maintenance, cell polarization, neoplastic invasiveness and metastasis, and membrane transport. It is suggested that Ksp-cadherin is a marker for terminal differentiation of the basolateral membranes of renal tubular epithelial cells. Within the kidney, Ksp-Cadherin is found exclusively in the basolateral membrane of renal tubular epithelial cells and collecting duct cells, and not in glomeruli, renal interstitial cells, or blood vessels. Ksp-Cadherin has been suggested to distinguish Chromophobe Renal-Cell Carcinoma from Oncocytoma.

Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 1 - 2 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) Optimal dilution of the Cadherin 16 antibody to be determined by the researcher.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM Tris with 1mM EDTA, pH 9.0, for 10 - 20 min followed by cooling at RT for 20 min.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: CDH16
Clonality: Monoclonal
Clone: CDH16/1071
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Mouse, Rat, Rabbit, Dog