Anti-SLIT2 Rabbit Polyclonal Antibody
PRSI16-009
New Product
- Antibody type:Primary
- Antigen name:Slit homolog 2 (Drosophila)
- Antigen symbol:SLIT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:9353
- Antigen synonyms:Slit-2|SLIL3|slit homolog 2 (Drosophila)|HGNC:11086|SLIT2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1330-1529 of human SLIT2 (NP_004778.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: SLIT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human