Anti-PFDN4 Rabbit Polyclonal Antibody
Catalog # PRSI15-973
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Prefoldin subunit 4
- Antigen symbol:PFDN4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:5203
- Antigen synonyms:C1|PFD4|prefoldin 4|prefoldin subunit 4|protein C-1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-134 of human PFDN4 (NP_002614.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PFDN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat