Anti-HSPA1A Mouse Monoclonal Antibody [clone: 1B10F3]
PRSI15-858
- Antibody type:Primary
- Antigen name:Heat shock 70 kDa protein 1A
- Antigen symbol:HSPA1A
- Clonality:Monoclonal
- Clone:[1B10F3]
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:3303
- Antigen synonyms:HSPA1|HSP70-1A|HSP72|HSPA1A|HSP70-1|HEL-S-103|HSP70I
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide of human HSPA1A
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: HSPA1A
Clonality: Monoclonal
Clone: [1B10F3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat