Anti-COL4A1 Mouse Monoclonal Antibody clone: [2F11] (Biotinylated)
Catalog # PRSI98-529
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:collagen type IV alpha 1 chain
- Antigen symbol:COL4A1
- Clonality:Monoclonal
- Clone:[2F11]
- Conjugation:Biotin
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:1282;1284;1285;1286;1287;1288
- Antigen synonyms:COLIV
- Modification:None
- Storage buffer:Supplied in PBS/NaN₃
- Storage temperature:Store vial at 2...8 °C
- Concentration:0,5 mg/ml
- Shipping temperature:20
- Immunogen:Native human type IV collagen
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: COL4A1
Clonality: Monoclonal
Clone: [2F11]
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human