Anti-DAAM1 Rabbit Polyclonal Antibody
Catalogus # PRSI14-704
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:dishevelled associated activator of morphogenesis 1
- Antigen symbol:DAAM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:23002
- Antigen synonyms:DAAM1|KIAA0666|FLJ41657
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 769-1068 of human DAAM1 (NP_001257449.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: DAAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat