Anti-HARBI1 Rabbit Polyclonal Antibody
Catalogus # PRSI14-676
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:harbinger transposase derived 1
- Antigen symbol:HARBI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:283254
- Antigen synonyms:harbinger transposase-derived nuclease|C11orf77|putative nuclease HARBI1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-349 of human HARBI1 (NP_776172.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: HARBI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse