Anti-USP24 Rabbit Polyclonal Antibody
PRSI13-739
New Product
- Antibody type:Primary
- Antigen name:ubiquitin specific peptidase 24
- Antigen symbol:USP24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:23358
- Antigen synonyms:ubiquitin thiolesterase 24|ubiquitin thioesterase 24|ubiquitin specific protease 24|deubiquitinating enzyme 24|ubiquitin-specific processing protease 24|ubiquitin carboxyl-terminal hydrolase 24
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 2431-2620 of human USP24 (NP_056121.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: USP24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat