Anti-RFTN1 Rabbit Polyclonal Antibody
Catalog # PRSI13-644
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:raftlin, lipid raft linker 1
- Antigen symbol:RFTN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:23180
- Antigen synonyms:MIG2|raft-linking protein|PIB1PIG9|proliferation-inducing protein 1proliferation-inducing protein 9|RAFTLIN|raftlin|cell migration-inducing gene 2 protein
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human RFTN1 (NP_055965.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RFTN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat