Anti-BCKDK Rabbit Polyclonal Antibody
Catalogus # PRSI62-669
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:[3-methyl-2-oxobutanoate dehydrogenase [lipoamide]] kinase, mitochondrial
- Antigen symbol:BCKDK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:120-151 aa
- Form:Liquid
- Gene ID:10295
- Modification:None
- Molecular weight:46 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This BCKDK antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 120-151 amino acids from the central region of human BCKDK.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.
Type: Primary
Antigen: BCKDK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 120-151
Host: Rabbit
Isotype: Ig
Reactivity: Human