Anti-PRKCG Rabbit Polyclonal Antibody
PRSI62-595
New Product
- Antibody type:Primary
- Antigen name:Protein kinase C gamma type
- Antigen symbol:PRKCG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Epitope:305-336 aa
- Form:Liquid
- Gene ID:5582
- Modification:None
- Molecular weight:78 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This PKC gamma antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 305-336 amino acids from the C-terminal region of human PKC gamma.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Pk:400 µl
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50.
Type: Primary
Antigen: PRKCG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 305-336
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse