Anti-CNGA2 Rabbit Polyclonal Antibody
PRSI61-685
New Product
- Antibody type:Primary
- Antigen name:Cyclic nucleotide-gated olfactory channel
- Antigen symbol:CNGA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:75-102 aa
- Form:Liquid
- Gene ID:1260
- Modification:None
- Molecular weight:76 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:This CNGA2 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 75-102 amino acids from the N-terminal region of human CNGA2.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: CNGA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 75-102
Host: Rabbit
Isotype: Ig
Reactivity: Human