Anti-EREG Rabbit Polyclonal Antibody
Catalog # PRSI61-668
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Proepiregulin
- Antigen symbol:EREG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:137-165 aa
- Form:Liquid
- Gene ID:2069
- Modification:None
- Molecular weight:19 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,45 mg/ml
- Shipping temperature:4 °C
- Immunogen:This EREG antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 137-165 amino acids from the C-terminal region of human EREG.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For FACS starting dilution is: 1:25. For IHC-P starting dilution is: 1:25. For WB starting dilution is: 1:1000.
Type: Primary
Antigen: EREG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 137-165
Host: Rabbit
Isotype: Ig
Reactivity: Human