Anti-NFYB Rabbit Polyclonal Antibody
Catalog # PRSI22-843
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Nuclear transcription factor Y, beta
- Antigen symbol:NFYB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:4801
- Antigen synonyms:CBF-A|HAP3|CBF-B|NFYB|NF-YB
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-207 of human NFYB (NP_006157.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: NFYB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse