Anti-VPS4A Rabbit Polyclonal Antibody
PRSI22-694
New Product
- Antibody type:Primary
- Antigen name:Vacuolar protein sorting 4 homolog A (S. cerevisiae)
- Antigen symbol:VPS4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:27183
- Antigen synonyms:VPS4|FLJ22197|VPS4-1|SKD2|SKD1A|SKD1|VPS4A
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human VPS4A (NP_037377.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: VPS4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat