Anti-MATK Rabbit Polyclonal Antibody
PRSI22-380
New Product
- Antibody type:Primary
- Antigen name:Megakaryocyte-associated tyrosine kinase
- Antigen symbol:MATK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:4145
- Antigen synonyms:HYLTK|CSK homologous kinase|hydroxyaryl-protein kinase|CTK|leukocyte carboxyl-terminal src kinase related|HHYLTK|megakaryocyte-associated tyrosine-protein kinase|tyrosine-protein kinase CTK|Lsk|protein kinase HYL|hematopoietic consensus tyrosine-lacking kinase|tyrosine kinase MATK|Csk-homologous kinase|tyrosylprotein kinase|CHK|Csk-type protein tyrosine kinase|HYL tyrosine kinase|HYL
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 352-466 of human MATK (NP_647611.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: MATK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat