Anti-SLC5A6 Rabbit Polyclonal Antibody
PRSI22-219
New Product
- Antibody type:Primary
- Antigen name:Solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6
- Antigen symbol:SLC5A6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:8884
- Antigen synonyms:SLC5A6|SMVT
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 500 to the C-terminus of human SLC5A6 (NP_066918.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: SLC5A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human