Anti-SLC26A4 Rabbit Polyclonal Antibody
Catalog # PRSI16-594
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Solute carrier family 26 member 4
- Antigen symbol:SLC26A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:5172
- Antigen synonyms:TDH2B|truncated solute carrier family 26|EVA|member 4|PDS|DFNB4|sodium-independent chloride/iodide transporter|pendrin|solute carrier family 26 (anion exchanger)
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 250-350 of human SLC26A4 (NP_000432,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SLC26A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse