Anti-GPR62 Rabbit Polyclonal Antibody
Catalogus # PRSI16-466
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:G protein-coupled receptor 62
- Antigen symbol:GPR62
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:118442
- Antigen synonyms:G protein-coupled receptor 62|KPG_005|GPR62|GPCR8|HGPCR8
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 150-250 of human GPR62 (NP_543141,3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GPR62
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat