Anti-TSFM Rabbit Polyclonal Antibody
Catalog # PRSI16-425
Leverancier: ProSci Inc.
Specificaties
- Antibody type:Primary
- Antigen name:Ts translation elongation factor, mitochondrial
- Antigen symbol:TSFM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:10102
- Antigen synonyms:EF-Tsmt|COXPD3|TSFM|EFTSMT|EFTS|EF-TS
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 245-346 of human TSFM (NP_001166167)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specificaties
Over dit artikel
Type: Primary
Antigen: TSFM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat