Anti-ATP10A Rabbit Polyclonal Antibody
PRSI16-291
- Antibody type:Primary
- Antigen name:ATPase phospholipid transporting 10A (putative)
- Antigen symbol:ATP10A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:57194
- Antigen synonyms:P4-ATPase flippase complex alpha subunit ATP10A|ATP10C|class V|type 10C|phospholipid-transporting ATPase VA|ATPVC|Class V|phospholipid transporting (P-type)|ATPase type IV|aminophospholipid translocase VA|ATPase|type 10A|ATPVA|probable phospholipid-transporting ATPase VA
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 440-690 of human ATP10A (NP_077816,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: ATP10A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human