Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-AGT Rabbit Polyclonal Antibody
  PRSI16-155
New Product
Anti-AGT Rabbit Polyclonal Antibody
  PRSI16-155
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Angiotensinogen
  • Antigen symbol:
    AGT
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    183
  • Antigen synonyms:
    Ang IV|Ang II|Angiotensin 1-10|Angiotensin-3|Angiotensin 1-7|Angiotensin IV|Angiotensin 1-9|Angiotensin 1-4|Angiotensin I|Des-Asp[1]-angiotensin II|Angiotensin-2|Angiotensin 1-8|Angiotensin-4|Ang I|Angiotensinogen|Angiotensin 1-5|Angiotensin 2-8|Angiotensin-1|Angiotensin II|Angiotensin 3-8|Ang III|Serpin A8|Angiotensin III|SERPINA8|AGT
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 35-285 of human AGT (NP_000020,1)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

 

 

Type: Primary
Antigen: AGT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat