Anti-USP44 Rabbit Polyclonal Antibody
Catalog # PRSI16-092
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin specific peptidase 44
- Antigen symbol:USP44
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:84101
- Antigen synonyms:ubiquitin thioesterase 44|ubiquitin carboxyl-terminal hydrolase 44|deubiquitinating enzyme 44|ubiquitin-specific-processing protease 44|ubiquitin specific protease 44|ubiquitin thiolesterase 44
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 100-270 of human USP44 (NP_115523,2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: USP44
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat