Anti-MRTO4 Rabbit Polyclonal Antibody
PRSI59-305
New Product
- Antibody type:Primary
- Antigen name:mRNA turnover protein 4 homolog {ECO:0000250|UniProtKB:P33201}
- Antigen symbol:MRTO4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:46 - 72 aa
- Form:Liquid
- Gene ID:51154
- Modification:None
- Molecular weight:28 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This MRTO4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 46 - 72 amino acids from the N-terminal region of human MRTO4.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: MRTO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 46-72
Host: Rabbit
Isotype: Ig
Reactivity: Human