Anti-WBSCR17 Rabbit Polyclonal Antibody
PRSI56-889
New Product
- Antibody type:Primary
- Antigen name:Putative polypeptide N-acetylgalactosaminyltransferase-like protein 3
- Antigen symbol:WBSCR17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:Amino acid range = 411-440
- Form:Liquid
- Gene ID:Q6IS24
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:68 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This WBSCR17 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 411-440 amino acids from the C-terminal region of human WBSCR17
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
This gene encodes an N-acetylgalactosaminyltransferase, which has 97% sequence identity to the mouse protein. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. [provided by RefSeq].
- For WB starting dilution is: 1:1000
Type: Primary
Antigen: WBSCR17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Mouse